CAS RN 41317-10-6
CAS RN 41317-10-6
IUPAC Name:
5-(dimethylamino)-2-nitrosophenol hydrochloride
Synonyms:
5-(dimethylamino)-2-nitrosophenol hydrochloride
Molecular Weight (g/mol):
202.64
Molecular Formula:
C8H11ClN2O2
InChi Key:
WRIZNEWVHJJMHU-UHFFFAOYSA-N
SMILES:
Cl.CN(C)C1=CC(O)=C(C=C1)N=O
1
–
1
of
1
results
1
2-nitroso-5-dimethylaminophenol hydrochloride, TRC
CAS: 41317-10-6 Molecular Formula: C8H10N2O2 . HCl Molecular Weight (g/mol): 202.64 IUPAC Name: 5-(dimethylamino)-2-nitrosophenol;hydrochloride SMILES: Cl.CN(C)c1ccc(N=O)c(O)c1