missing translation for 'onlineSavingsMsg'
Learn More

2-nitroso-5-dimethylaminophenol hydrochloride, TRC

Product Code. 30472386
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30472386 500 mg
30465647 100 mg
30464342 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472386 Supplier Toronto Research Chemicals Supplier No. TRCN497233500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41317-10-6
Molecular Formula C8H10N2O2 . HCl
Molecular Weight (g/mol) 202.64
IUPAC Name 5-(dimethylamino)-2-nitrosophenol;hydrochloride
SMILES Cl.CN(C)c1ccc(N=O)c(O)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H10N2O2.ClH/c1-10(2)6-3-4-7(9-12)8(11)5-6;/h3-5,11H,1-2H3;1H
Formula Weight 202.0509
Chemical Name or Material 2-nitroso-5-dimethylaminophenol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.