accessibility menu, dialog, popup

UserName

BrO

CAS RN 4133-35-1

IUPAC Name: 6-bromo-1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 6-bromo-3,4-dihydro-1H-naphthalen-2-one
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: BYHKDUFPSJWJDI-UHFFFAOYSA-N
SMILES: BrC1=CC=C2CC(=O)CCC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results