accessibility menu, dialog, popup

UserName

ClOS

CAS RN 41507-35-1

IUPAC Name: thiophene-3-carbonyl chloride
Synonyms: thiophene-3-carbonyl chloride
Molecular Weight (g/mol): 146.59
Molecular Formula: C5H3ClOS
InChi Key: QTWBEVAYYDZLQL-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results