missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15487968
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487968 25 g 25g
15477968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487968 Supplier Thermo Scientific Alfa Aesar Supplier No. H61428.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41507-35-1
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00130090
InChI Key QTWBEVAYYDZLQL-UHFFFAOYSA-N
Synonym 3-thiophenecarbonyl chloride, 3-thenoyl chloride, 3-thiophenecarboxylic acid chloride, 3-thiophenecarbonylchloride, pubchem5597, 3-chlorocarbonyl thiophene, 3-thiophene carbonyl chloride, thiophene-3-carboxylic acid chloride, thiophene-3-carbonyl chloride, 3-thiophenecarbonyl chloride 6ci, 9ci
PubChem CID 2776377
IUPAC Name thiophene-3-carbonyl chloride
SMILES ClC(=O)C1=CSC=C1

Specifications

Melting Point 51°C to 54°C
Flash Point 51°C (124°F)
Quantity 25 g
UN Number UN2921
Sensitivity Moisture sensitive
Formula Weight 146.59
Percent Purity 97%
Chemical Name or Material Thiophene-3-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.