accessibility menu, dialog, popup

UserName

CAS RN 4160-61-6

IUPAC Name: methyl 4-oxothiane-3-carboxylate
Synonyms: methyl 4-oxothiane-3-carboxylate
Molecular Weight (g/mol): 174.21
Molecular Formula: C7H10O3S
InChi Key: MCUXKFHPGMEIIW-UHFFFAOYNA-N
SMILES: COC(=O)C1CSCCC1=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results