missing translation for 'onlineSavingsMsg'
Learn More

Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate, 95%

Product Code. 10481440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10481440 5 g Glass bottle 5g
10352450 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10481440 Supplier Thermo Scientific Acros Supplier No. 434290050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4160-61-6 | C7H10O3S | 174.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4160-61-6
Molecular Formula C7H10O3S
Molecular Weight (g/mol) 174.22
InChI Key MCUXKFHPGMEIIW-UHFFFAOYSA-N
Synonym methyl 4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl tetrahydro-4-oxo-2h-thiopyran-3-carboxylate, methyl 4-oxotetrahydrothiopyran-3-carboxylate, 2h-thiopyran-3-carboxylic acid, tetrahydro-4-oxo-, methyl ester, methyl4-oxotetrahydro-2h-thiopyran-3-carboxylate, methyl-4-oxotetrahydro-2h-thiopyran-3-carboxylate, 2,3,5,6-tetrahydro-3-carbomethoxythiopyran-4-one, 4-oxotetrahydro-2h-thiopyran-3-carbonsauremethylester, 4-oxotetrahydro-2h-thiopyran-3-carboxylic acid methyl ester, 4-oxo-tetrahydro-thiopyran-3-carboxylic acid methyl ester
PubChem CID 10374916
IUPAC Name methyl 4-oxothiane-3-carboxylate
SMILES COC(=O)C1CSCCC1=O

Specifications

Color Colorless to Yellow
Boiling Point 134°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 174.22
Percent Purity 95%
Physical Form Liquid or Solid
Chemical Name or Material Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
compliance-icons
Product Identifier
  • Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.