accessibility menu, dialog, popup

UserName

CAS RN 4170-30-3

CH3O(Z)

CAS RN 4170-30-3

IUPAC Name: (2Z)-but-2-enal
Synonyms: crotonaldehyde
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: MLUCVPSAIODCQM-IHWYPQMZSA-N
SMILES: C\C=C/C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results