missing translation for 'onlineSavingsMsg'
Learn More

Crotonaldehyde, 99+%

Product Code. 10327480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327480 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10327480 Supplier Thermo Scientific Acros Supplier No. 158220010
Quantity Price
1 197.11 € / 1 L
6 181.20 € / 1 L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 4170-30-3 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4170-30-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00007003
InChI Key MLUCVPSAIODCQM-NSCUHMNNSA-N
Synonym crotonaldehyde, 2-butenal, crotylaldehyde, crotonal, 2e-but-2-enal, trans-crotonaldehyde, e-crotonaldehyde, crotonic aldehyde, 1-formylpropene, trans-2-butenal
PubChem CID 447466
ChEBI CHEBI:41607
IUPAC Name (E)-but-2-enal
SMILES CC=CC=O

Specifications

Melting Point -74.0°C
Density 0.8580g/mL
Boiling Point 101.0°C to 103.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCHO
Refractive Index 1.4355 to 1.4375
Quantity 1 L
Beilstein , 728
Merck Index 15, 2587
Specific Gravity 0.858
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,toluene and,kerosene,miscible in all proportions with gasoline,solvent,very soluble in acetone
Viscosity 0.27 mPa.s (20°C)
Formula Weight 70.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Crotonaldehyde, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Crotonaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.