accessibility menu, dialog, popup

UserName

CH3CH2OPOOH3COH3C

CAS RN 4185-82-4

IUPAC Name: dimethyl prop-1-en-2-yl phosphate
Synonyms: dimethyl prop-1-en-2-yl phosphate
Molecular Weight (g/mol): 166.11
Molecular Formula: C5H11O4P
InChi Key: CLUBBNMVLUHMDR-UHFFFAOYSA-N
SMILES: COP(=O)(OC)OC(C)=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results