missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl isopropenyl phosphate, 95%, Thermo Scientific™

Product Code. 11377109
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377109 100 g 100g
11357109 5 g 5g
11367109 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377109 Supplier Thermo Scientific Alfa Aesar Supplier No. H27353.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4185-82-4
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.113
MDL Number MFCD09953485
InChI Key CLUBBNMVLUHMDR-UHFFFAOYSA-N
Synonym dimethyl isopropenyl phosphate, phosphoric acid, isopropenyl dimethyl ester, phosphoric acid, dimethyl 1-methylethenyl ester, isopropenyl dimethyl phosphate, 1-propen-2-yl-dimethyl phosphate, dimethyl 1-methylethenyl phosphate, phosphoric acid dimethyl=1-methylvinyl ester, isopropenyl methyl phosphate, c3h5o meo 2po
PubChem CID 581450
IUPAC Name dimethyl prop-1-en-2-yl phosphate
SMILES CC(=C)OP(=O)(OC)OC

Specifications

Boiling Point 120°C (7.5 mmHg)
Refractive Index 1.42
Quantity 100 g
Sensitivity Moisture sensitive
Formula Weight 166.11
Percent Purity 95%
Chemical Name or Material Dimethyl isopropenyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.