accessibility menu, dialog, popup

UserName

OHHC

CAS RN 4187-87-5

IUPAC Name: 1-phenylprop-2-yn-1-ol
Synonyms: benzenemethanol, α-ethynyl-
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: UIGLAZDLBZDVBL-UHFFFAOYNA-N
SMILES: OC(C#C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)