missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propyn-1-ol, 98%

Product Code. 11357486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357486 5 g 5g
11347486 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357486 Supplier Thermo Scientific Alfa Aesar Supplier No. L09549.06
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4187-87-5 | C9H8O | 132.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4187-87-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00021860
InChI Key UIGLAZDLBZDVBL-UHFFFAOYSA-N
Synonym 1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
PubChem CID 20155
IUPAC Name 1-phenylprop-2-yn-1-ol
SMILES C#CC(C1=CC=CC=C1)O

Specifications

Melting Point 26°C to 30°C
Density 1.087
Boiling Point 231°C to 233°C
Flash Point 99°C (210°F)
Refractive Index 1.55
Quantity 5 g
Beilstein 742365
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material 1-Phenyl-2-propyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.