accessibility menu, dialog, popup

UserName

CAS RN 42189-56-0

IUPAC Name: 1-(pyren-4-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(pyren-4-yl)pyrrole-2,5-dione
Molecular Weight (g/mol): 297.31
Molecular Formula: C20H11NO2
InChi Key: NPTUGEKDRBZJRE-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC2=CC=CC3=CC=C4C=CC=C1C4=C23
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results