accessibility menu, dialog, popup

UserName

CAS RN 422-64-0

IUPAC Name: silver(1+) pentafluoropropanoate
Synonyms: silver(1+) pentafluoropropanoate
Molecular Weight (g/mol): 270.89
Molecular Formula: C3AgF5O2
InChi Key: XAYJXAUUXJTOSI-UHFFFAOYSA-M
SMILES: [Ag+].[O-]C(=O)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)