accessibility menu, dialog, popup

UserName

H3CCH3HO

CAS RN 4221-99-2

IUPAC Name: butan-2-ol
Synonyms: 2-butanol butanol secondaire s-butanol 2-butanol, (-)-isomer epa pesticide chemical code 001502 2-butanol, (+)-isomer (+-)-2-butanol 2-butanol, (+-)- s-butyl alcohol (+)-2-butanol
Molecular Weight (g/mol): 74.12
Molecular Formula: C4H10O
InChi Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILES: CCC(C)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results