missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-sec-Butanol, 99%

Product Code. 10388350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10388350 5 g 5g
10022990 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388350 Supplier Thermo Scientific Acros Supplier No. 218030050
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4221-99-2 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4221-99-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00064281
InChI Key BTANRVKWQNVYAZ-BYPYZUCNSA-N
Synonym s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
PubChem CID 444683
ChEBI CHEBI:45475
IUPAC Name (2S)-butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -114°C
CAS Min % 98.5
Color Colorless
CAS Max % 100
Density 0.8000g/mL
Quantity 5 g
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.398
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.8
Linear Formula C2H5CH(OH)CH3
Viscosity 4.21 mPa.s (20°C)
Beilstein 01, 373
Merck Index 15, 1543
Boiling Point 99°C to 100°C
Solubility Information Solubility in water: in water: 125g/L (20°C)
Formula Weight 74.12
Percent Purity 99%
Chemical Name or Material (S)-(+)-sec-Butanol, 0.99
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-sec-Butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.