accessibility menu, dialog, popup

UserName

CH3H3CCH3NH2Cl

CAS RN 42265-67-8

IUPAC Name: 4-tert-butyl-2-chloroaniline
Synonyms: 4-tert-butyl-2-chloroaniline
Molecular Weight (g/mol): 183.68
Molecular Formula: C10H14ClN
InChi Key: MFEZEFHCSBXPPO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Cl)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results