accessibility menu, dialog, popup

UserName

CAS RN 4230-93-7

IUPAC Name: 1,2-dimethoxy-4-[(1E)-2-nitroethenyl]benzene
Synonyms: 1,2-dimethoxy-4-[(1E)-2-nitroethenyl]benzene
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: SYJMYDMKPSZMSB-AATRIKPKSA-N
SMILES: COC1=CC=C(\C=C\[N+]([O-])=O)C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results