accessibility menu, dialog, popup

UserName

CAS RN 423118-47-2

BCH3CH3H3CFFFK

CAS RN 423118-47-2

IUPAC Name: potassium (4-tert-butylphenyl)trifluoroboranuide
Synonyms: potassium (4-tert-butylphenyl)trifluoroboranuide
Molecular Weight (g/mol): 240.12
Molecular Formula: C10H13BF3K
InChi Key: RQPOMNZPBPIWDB-UHFFFAOYSA-N
SMILES: [K+].CC(C)(C)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results