CAS RN 423768-50-7
IUPAC Name:
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
Synonyms:
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol):
302.14
Molecular Formula:
C10H8BrNO3S
InChi Key:
OIZZMDOBYXRSNQ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NOC(=C1)C1=CC=C(Br)S1
1
–
1
of
1
results
1
ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™
CAS: 423768-50-7 Molecular Formula: C10H8BrNO3S Molecular Weight (g/mol): 302.142 MDL Number: MFCD03407336 InChI Key: OIZZMDOBYXRSNQ-UHFFFAOYSA-N Synonym: ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate,ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate,ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate PubChem CID: 2776548 IUPAC Name: ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate SMILES: CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br