missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Product Code. 10495312
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10495312 250 mg 250mg
10140832 1 g 1g
10419084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10495312 Supplier Thermo Scientific Maybridge Supplier No. CC23210CB
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 423768-50-7
Molecular Formula C10H8BrNO3S
Molecular Weight (g/mol) 302.142
MDL Number MFCD03407336
InChI Key OIZZMDOBYXRSNQ-UHFFFAOYSA-N
Synonym ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate
PubChem CID 2776548
IUPAC Name ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 302.15
Percent Purity 97%
Chemical Name or Material ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.