accessibility menu, dialog, popup

UserName

H3CSNNNH2

CAS RN 42382-46-7

IUPAC Name: 2-(methylsulfanyl)pyrimidin-5-amine
Synonyms: 2-(methylsulfanyl)pyrimidin-5-amine
Molecular Weight (g/mol): 141.19
Molecular Formula: C5H7N3S
InChi Key: NRJNAXLXHFQYEX-UHFFFAOYSA-N
SMILES: CSC1=NC=C(N)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results