missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(methylthio)pyrimidine, 96%, Thermo Scientific™

Product Code. 15535886 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535886 250 mg 250mg
15525886 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535886 Supplier Thermo Scientific Acros Supplier No. 449822500
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 42382-46-7
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.19
MDL Number MFCD10697095
InChI Key NRJNAXLXHFQYEX-UHFFFAOYSA-N
Synonym 5-amino-2-methylthio pyrimidine, 2-methylthio pyrimidin-5-amine, 2-methylsulfanyl pyrimidin-5-amine, 2-methylmercapto-5-aminopyrimidine, 5-pyrimidinamine,2-methylthio, 2-methylsulfanylpyrimidin-5-ylamine, 2-methylsulfanyl-5-pyrimidinamine, 2-methylthio-5-pyrimidinamine, 5-pyrimidinamine, 2-methylthio, 5-pyrimidinamine, 2-methylthio-9ci
PubChem CID 12215991
IUPAC Name 2-methylsulfanylpyrimidin-5-amine
SMILES CSC1=NC=C(N)C=N1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Formula Weight 141.19
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(methylthio)pyrimidine
compliance-icons
Product Identifier
  • 5-Amino-2-(methylthio)pyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.