CAS RN 42397-65-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,8-Dinitropyrene (90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,8-Dinitropyrene (90%), TRC
CAS: 42397-65-9 Molecular Formula: C16 H8 N2 O4 Molecular Weight (g/mol): 292.25 Synonym: Pyrene, 1,8-dinitro-,1,8-Dinitropyrene IUPAC Name: 1,8-dinitropyrene SMILES: [O-][N+](=O)c1ccc2ccc3ccc(c4ccc1c2c34)[N+](=O)[O-]