accessibility menu, dialog, popup

UserName

CAS RN 424-40-8

CH3OOFFFFFFOOH3C

CAS RN 424-40-8

IUPAC Name: 1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Synonyms: 1,5-diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Molecular Weight (g/mol): 296.17
Molecular Formula: C9H10F6O4
InChi Key: MSDPXVBLFJODJO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results