missing translation for 'onlineSavingsMsg'
Learn More

Diethyl hexafluoroglutarate, 97%, Thermo Scientific Chemicals

Product Code. 11430567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430567 5 g 5g
11420567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430567 Supplier Thermo Scientific Alfa Aesar Supplier No. B22428.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 424-40-8
Molecular Formula C9H10F6O4
Molecular Weight (g/mol) 296.17
MDL Number MFCD00015156
InChI Key MSDPXVBLFJODJO-UHFFFAOYSA-N
Synonym diethyl hexafluoroglutarate, diethyl perfluoroglutarate, diethyl hexafluoropentanedioate, hexafluoroglutaric acid diethyl ester, pentanedioic acid, hexafluoro-, diethyl ester, glutaric acid, hexafluoro-, diethyl ester, hexafluoropentanedioic acid diethyl ester, diethylester kyseliny hexafluorglutarove, diethylester kyseliny hexafluorglutarove czech, pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester
PubChem CID 9874
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OCC

Specifications

Density 1.365
Boiling Point 224°C
Refractive Index 1.36
Quantity 5 g
Beilstein 1804802
IUPAC Name diethyl 2,2,3,3,4,4-hexafluoropentanedioate
Formula Weight 296.16
Percent Purity 97%
Chemical Name or Material Diethyl hexafluoroglutarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.