accessibility menu, dialog, popup

UserName

CAS RN 42518-98-9

SClOCl

CAS RN 42518-98-9

IUPAC Name: 5-chlorothiophene-2-carbonyl chloride
Synonyms: 5-chlorothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H2Cl2OS
InChi Key: BMPDCQVRKDNUAP-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(Cl)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results