missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carbonyl chloride, 98%

Product Code. 15498018
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498018 5 g 5g
15408028 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498018 Supplier Thermo Scientific Alfa Aesar Supplier No. H61474.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42518-98-9 | C5H2Cl2OS | 181.03 g/mol

5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorothiophene-2-carbonyl chloride used as a intermediate for rivaroxaban.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool, dry place in tightly closed container. Store away form water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42518-98-9
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00043886
InChI Key BMPDCQVRKDNUAP-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-carbonylchloride, 5-chorothiophene-2-carbonyl chloride, 5-chloro-2-thiophenecarbonyl chloride, 5-chloro-thiophene-2-carbonyl chloride, 5-chloro-2-thenoyl chloride, 2-thiophenecarbonyl chloride, 5-chloro, pubchem16132, ksc235i7t, 5-chlorothiophenecarbonyl chloride, 5-chloro-2-thenoylchloride
PubChem CID 589223
IUPAC Name 5-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 4°C
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.