accessibility menu, dialog, popup

UserName

BrSOOONa

CAS RN 4263-52-9

IUPAC Name: sodium 2-bromoethane-1-sulfonate
Synonyms: sodium 2-bromoethanesulfonate
Molecular Weight (g/mol): 211
Molecular Formula: C2H4BrNaO3S
InChi Key: HNFOAHXBHLWKNF-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCBr
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)