missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanesulfonic Acid Sodium Salt, TRC

Product Code. 30344903
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344903 100 g 100g
30332023 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30344903 Supplier Toronto Research Chemicals Supplier No. TRCB684290100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoethanesulfonic Acid Sodium Salt
CAS 4263-52-9
Molecular Formula C2H4BrNaO3S
Quantity 100 g
Synonym Sodium 2-Bromoethanesulfonate, Sodium Bromoethylsulfonate, 2-Bromoethanesulfonic Acid Sodium Salt
InChI Formula InChI=1S/C2H5BrO3S.Na/c3-1-2-7(4,5)6;/h1-2H2,(H,4,5,6);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)CCBr
IUPAC Name sodium;2-bromoethanesulfonate
Molecular Weight (g/mol) 211.01
Formula Weight 209.8962
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.