accessibility menu, dialog, popup

UserName

CAS RN 4282-40-0

H3CI

CAS RN 4282-40-0

IUPAC Name: 1-iodoheptane
Synonyms: 1-iodoheptane
Molecular Weight (g/mol): 226.1
Molecular Formula: C7H15I
InChi Key: LMHCYRULPLGEEZ-UHFFFAOYSA-N
SMILES: CCCCCCCI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)