missing translation for 'onlineSavingsMsg'
Learn More

1-Iodoheptane, 98%, stab. with copper

Product Code. 11399829
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11399829

Brand: Thermo Scientific Alfa Aesar B24206.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4282-40-0 | C7H15I | 226.101 g/mol

1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4282-40-0
Molecular Formula C7H15I
Molecular Weight (g/mol) 226.101
MDL Number MFCD00001104
InChI Key LMHCYRULPLGEEZ-UHFFFAOYSA-N
Synonym heptyl iodide, n-heptyl iodide, heptane, 1-iodo, iodoheptane, n-heptyliodide, iodo heptane, 1-iodanylheptane, 1-iodomeptane, 1-iodo heptane, 1-iodo-heptane
PubChem CID 20274
IUPAC Name 1-iodoheptane
SMILES CCCCCCCI

Specifications

Melting Point -48°C
Density 1.379
Boiling Point 203°C to 205°C
Flash Point 78°C (174°F)
Refractive Index 1.49
Quantity 250 g
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 226.1
Percent Purity 98%
Chemical Name or Material 1-Iodoheptane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.