accessibility menu, dialog, popup

UserName

OFFFClOOH

CAS RN 433926-46-6

IUPAC Name: 3-chloro-5-(trifluoromethoxy)benzoic acid
Synonyms: 3-chloro-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 240.56
Molecular Formula: C8H4ClF3O3
InChi Key: KVXQRRFQLRUCKN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OC(F)(F)F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results