accessibility menu, dialog, popup

UserName

CH3H3COH

CAS RN 4383-18-0

IUPAC Name: 2-phenylpentan-2-ol
Synonyms: 2-phenylpentan-2-ol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: YBQOTTAEVBHNEJ-UHFFFAOYNA-N
SMILES: CCCC(C)(O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results