missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-pentanol, 98%

Product Code. 15423227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423227 1 g 1g
11435806 5 g 5g
11445806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423227 Supplier Thermo Scientific Alfa Aesar Supplier No. B20854.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4383-18-0 | C11H16O | 164.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4383-18-0
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00046592
InChI Key YBQOTTAEVBHNEJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-pentanol, 2-phenyl-pentan-2-ol, benzenemethanol, .alpha.-methyl-.alpha.-propyl, methyl phenylbutanol, acmc-1aei8, methyl phenyl n-propyl carbinol
PubChem CID 138214
IUPAC Name 2-phenylpentan-2-ol
SMILES CCCC(C)(C1=CC=CC=C1)O

Specifications

Quantity 1 g
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 2-Phenyl-2-pentanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.