accessibility menu, dialog, popup

UserName

CAS RN 4386-39-4

CH3OOCH3OHO

CAS RN 4386-39-4

IUPAC Name: 2-(acetyloxy)-3-methylbenzoic acid
Synonyms: 2-(acetyloxy)-3-methylbenzoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: XRBMKGUDDJPAMH-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(C)C=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results