accessibility menu, dialog, popup

UserName

CAS RN 4410-99-5

IUPAC Name: 2-phenylethane-1-thiol
Synonyms: 2-phenylethanethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: ZMRFRBHYXOQLDK-UHFFFAOYSA-N
SMILES: SCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results