missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanethiol, 99%

Product Code. 10769383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10769383 10 g Glass bottle 10g
10520764 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10769383 Supplier Thermo Scientific Acros Supplier No. 378490100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4410-99-5 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4410-99-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00004891
InChI Key ZMRFRBHYXOQLDK-UHFFFAOYSA-N
Synonym phenethyl mercaptan, benzeneethanethiol, 2-phenylethyl mercaptan, phenylethyl mercaptan, 2-phenylethylthiol, unii-i4th2t2iny, 2-phenyl-ethanethiol, 2-phenylethylmercaptan, beta-phenylethyl mercaptan, i4th2t2iny
PubChem CID 78126
SMILES SCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0300g/mL
Boiling Point 97°C to 100°C (12.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5(CH2)2SH
Refractive Index 1.5590 to 1.561
Quantity 10 g
Specific Gravity 1.03
IUPAC Name 2-phenylethane-1-thiol
Formula Weight 138.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Phenylethanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.