CAS RN 441045-17-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eribulin mesylate, MedChemExpress
MedChemExpress Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used for the research of metastatic breast cancer. Eribulin mesylate inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules.