missing translation for 'onlineSavingsMsg'
Learn More

Eribulin mesylate, MedChemExpress

Product Code. 16441349
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441349 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441349 Supplier Medchem Express Supplier No. HY13442A
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used for the research of metastatic breast cancer. Eribulin mesylate inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules.

Eribulin mesylate (E7389 mesylate) is a microtubule targeting agent that is used for the research of metastatic breast cancer. Eribulin mesylate inhibits the proliferation of cancer cells by binding microtubule proteins and microtubules.

Specifications

Chemical Name or Material Eribulin mesylate
CAS 441045-17-6
Color White
Purity Grade Notes Research
Molecular Formula C41H63NO14S
Quantity 1 mg
Synonym B1939 mesylateE7389 mesylateER-086526 mesylate
Solubility Information DMSO : ≥ 100 mg/mL (121.07 mM) ∣Ethanol : ≥ 100 mg/mL (121.07 mM)
SMILES C=C1C[C@@](CC[C@@]2(C[C@@]3([H])O4)O[C@]([C@](O[C@](C5)([H])CC6)([H])[C@@]6([H])O7)([H])[C@@]4([H])[C@]7([H])[C@@]3([H])O2)([H])O[C@@]1([H])CC[C@](C[C@@H](C)C8=C)([H])O[C@]8([H])C[C@@](O[C@H](C[C@H](O)CN)[C@@H]9OC)([H])[C@]9([H])CC5=O.CS(=O)(O)=O
Molecular Weight (g/mol) 826.0
Formula Weight 826.0
Percent Purity 95.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.