accessibility menu, dialog, popup

UserName

CH3FF

CAS RN 443-84-5

IUPAC Name: 1,3-difluoro-2-methylbenzene
Synonyms: 1,3-difluoro-2-methylbenzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: MZLSNIREOQCDED-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results