CAS RN 443104-02-7
CAS RN 443104-02-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
CDDO Imidazolide, TRC
CAS: 443104-02-7 Molecular Formula: C34H43N3O3 Molecular Weight (g/mol): 541.72 Synonym: 1-(2-Cyano-3,12,28-trioxooleana-1,9(11)-dien-28-yl)-1H-imidazole,2-Cyano-3,12-dioxooleana-1,9(11)-dien-28-oic acid imidazolide,CDDO-Im,RTA 403,TP 235 IUPAC Name: (4aR,6aR,6aS,6bR,8aS,12aS,14bS)-8a-(imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-4a,5,6,6a,7,8,9,10,12,12a-decahydropicene-2-carbonitrile SMILES: CC1(C)CC[C@@]2(CC[C@]3(C)[C@@H]([C@@H]2C1)C(=O)C=C4[C@@]5(C)C=C(C#N)C(=O)C(C)(C)[C@@H]5CC[C@]43C)C(=O)N6C=CN=C6
2
CDDO-Im, MedChemExpress
MedChemExpress CDDO-Im (RTA-403) is an activator of Nrf2 and PPAR, with Kis of 232 and 344 nM for PPARα and PPARγ.