CAS RN 445295-04-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CP671305, MedChemExpress
MedChemExpress CP671305 is a potent, orally active, selective inhibitor of phosphodiesterase-4-D, and possesses high activities.