accessibility menu, dialog, popup

UserName

NHONOONOOCl

CAS RN 446855-41-0

IUPAC Name: N-[(4-chlorophenyl)methyl]-3,5-dinitrobenzamide
Synonyms: N-[(4-chlorophenyl)methyl]-3,5-dinitrobenzamide
Molecular Weight (g/mol): 335.7
Molecular Formula: C14H10ClN3O5
InChi Key: ZDFCVPAIESTUKH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1)C(=O)NCC1=CC=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results