accessibility menu, dialog, popup

UserName

CH3NCS

CAS RN 4478-92-6

IUPAC Name: (1-isothiocyanatoethyl)benzene
Synonyms: (1-isothiocyanatoethyl)benzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: QQCJPTVZIZVKEZ-UHFFFAOYNA-N
SMILES: CC(N=C=S)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results