accessibility menu, dialog, popup

UserName

CAS RN 4480-83-5

OOOO

CAS RN 4480-83-5

IUPAC Name: 1,4-dioxane-2,6-dione
Synonyms: 1,4-dioxane-2,6-dione
Molecular Weight (g/mol): 116.07
Molecular Formula: C4H4O4
InChi Key: PIYNUZCGMLCXKJ-UHFFFAOYSA-N
SMILES: O=C1COCC(=O)O1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)