missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic anhydride, 97%

Product Code. 11425967
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425967 50 g 50g
11415967 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425967 Supplier Thermo Scientific Alfa Aesar Supplier No. B24498.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4480-83-5 | C4H4O4 | 116.072 g/mol

Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diglycolic anhydride is used in the preparation of N,N-diethyldiglycollamsaeure by reacting with diethylamine. It is an useful bio chemical for proteomics research.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4480-83-5
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.072
MDL Number MFCD00006677
InChI Key PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Synonym diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
PubChem CID 78232
IUPAC Name 1,4-dioxane-2,6-dione
SMILES C1C(=O)OC(=O)CO1

Specifications

Melting Point 91°C to 94°C
Boiling Point 240°C to 241°C
Quantity 50 g
Beilstein 112527
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 116.07
Percent Purity 0.97
Chemical Name or Material Diglycolic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.