accessibility menu, dialog, popup

UserName

H3COFO

CAS RN 450-83-9

IUPAC Name: 4-fluoro-2-methoxybenzaldehyde
Synonyms: 4-fluoro-2-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: PTKRQIRPNNIORO-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results