accessibility menu, dialog, popup

UserName

CAS RN 452-81-3

CH3OHF

CAS RN 452-81-3

IUPAC Name: 2-fluoro-4-methylphenol
Synonyms: 2-fluoro-4-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: WJKISRFVKIOBCQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results